Learn More
FABP6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13781-1-AP-20UL
This item is not returnable.
View return policy
Description
The FABP6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FABP6. This antibody recognizes human, mouse antigen. The FABP6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| FABP6 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2172 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |