missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP6 Rabbit anti-Human, Clone: 201, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | FABP6 |
|---|---|
| Clone | 201 |
| Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260527
|
Novus Biologicals
NBP2-90127-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18216516
|
Novus Biologicals
NBP2-90127-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP6 Monoclonal specifically detects FABP6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| FABP6 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15PIntestinal 15 kDa protein, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBPIleal lipid-binding protein, illeal lipid-binding protein | |
| Recombinant Human FABP6 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 201 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 2172 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title