missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FABP5 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870014
|
Proteintech
12348-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860014
|
Proteintech
12348-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fatty acid-binding proteins, designated FABPs, are a family of homologous cytoplasmic proteins that are expressed in a highly tissue-specific manner and play an integral role in the balance between lipid and carbohydrate metabolism. FABPs mediate fatty acid (FA) and/or hydrophobic ligand uptake, transport and targeting within their respective tissues. The mechanisms underlying these actions can give rise to both passive diffusional uptake and protein-mediated transmembrane transport of FAs. FABPs are expressed in adipocytes (A-FABP), brain (B-FABP), epidermis (E-FABP, also designated psoriasis-associated FABP or PA-FABP), muscle and heart (H-FABP, also designated mammary-derived growth inhibitor or MDGI), intestine (I-FABP), liver (L-FABP), myelin (M-FABP) and testis (T-FABP). Epithelial fatty acid-binding protein (E-FABP) binds stearic acid and may play a role in keratinocyte differentiation. E-FABP is upregulated in rat dorsal root ganglia after sciatic nerve crush and in differentiating neurons during development.Specifications
| FABP5 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P55053, Q01469, Q05816 | |
| 140868, 16592, 2171 | |
| FABP5 Fusion Protein Ag3005 | |
| Primary | |
| -20°C | |
| FABP5 |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| E FABP, EFABP, FABP5, Fatty acid binding protein 5, PA FABP, PAFABP | |
| FABP5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title