missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5 Mouse anti-Human, Clone: 1C6E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66299-1-IG-150UL
This item is not returnable.
View return policy
Description
Fatty acid-binding proteins, designated FABPs, are a family of homologous cytoplasmic proteins that are expressed in a highly tissue-specific manner and play an integral role in the balance between lipid and carbohydrate metabolism. FABPs mediate fatty acid (FA) and/or hydrophobic ligand uptake, transport and targeting within their respective tissues. The mechanisms underlying these actions can give rise to both passive diffusional uptake and protein-mediated transmembrane transport of FAs. FABPs are expressed in adipocytes (A-FABP), brain (B-FABP), epidermis (E-FABP, also designated psoriasis-associated FABP or PA-FABP), muscle and heart (H-FABP, also designated mammary-derived growth inhibitor or MDGI), intestine (I-FABP), liver (L-FABP), myelin (M-FABP) and testis (T-FABP). Epithelial fatty acid-binding protein (E-FABP) binds stearic acid and may play a role in keratinocyte differentiation. E-FABP is upregulated in rat dorsal root ganglia after sciatic nerve crush and in differentiating neurons during development.Specifications
| FABP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01469 | |
| FABP5 | |
| FABP5 Fusion Protein Ag3005 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1C6E12 | |
| Unconjugated | |
| FABP5 | |
| E FABP, EFABP, FABP5, Fatty acid binding protein 5, PA FABP, PAFABP | |
| Mouse | |
| Protein G | |
| RUO | |
| 2171 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction