missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5/E-FABP Rabbit anti-Human, Clone: FABP5/6353R, Novus Biologicals™
Rabbit Monoclonal Antibody
£228.00 - £411.00
Specifications
| Antigen | FABP5/E-FABP |
|---|---|
| Clone | FABP5/6353R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18395644
|
Novus Biologicals
NBP3-13882-20UG |
20 μg |
£228.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329906
|
Novus Biologicals
NBP3-13882-100UG |
100 μg |
£411.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP5/E-FABP Monoclonal antibody specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FABP5/E-FABP | |
| 0.2 mg/ml | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| Recombinant human full-length FABP5/E-FABP protein (Uniprot: Q01469) | |
| Primary | |
| Store at 4°C. |
| FABP5/6353R | |
| Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q01469 | |
| 2171 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title