missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5/E-FABP, Mouse anti-Human, Clone: FABP5/3750, Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £377.00
Specifications
| Antigen | FABP5/E-FABP |
|---|---|
| Clone | FABP5/3750 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 0.1-0.2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209887
|
Novus Biologicals
NBP2-79795-20UG |
20 μg |
£211.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286226
|
Novus Biologicals
NBP2-79795-100UG |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| FABP5/E-FABP | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| FABP5 | |
| IgG2a κ | |
| Protein A or G purified |
| FABP5/3750 | |
| Immunohistochemistry 0.1-0.2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q01469, Q01469 | |
| 2171 | |
| Recombinant full-length human FABP5/E-FABP protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title