missing translation for 'onlineSavingsMsg'
Learn More

FABP5/E-FABP Antibody (FABP5/6353R), HRP, Novus Biologicals™

Product Code. 18366875 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366875 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366875 Supplier Novus Biologicals Supplier No. NBP314216H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FABP5/E-FABP Monoclonal antibody specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP5/E-FABP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FABP5/6353R
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog
Host Species Rabbit
Immunogen Recombinant human full-length FABP5/E-FABP protein (Uniprot: Q01469)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2171
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.