Learn More
Invitrogen™ FABP4 Monoclonal Antibody (9B8D)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515566
Description
MA5-15566 targets FABP4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15566 immunogen is purified recombinant fragment of FABP4 expressed in E. Coli. MA5-15566 detects FABP4 which has a predicted molecular weight of approximately 14kDa.
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
Specifications
| FABP4 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P04117, P15090, P70623 | |
| FABP4 | |
| Purified recombinant fragment of FABP4 expressed in E. Coli. | |
| RUO | |
| 11770, 2167, 79451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 9B8D | |
| Unconjugated | |
| FABP4 | |
| 3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid binding protein; adipocyte lipid-binding protein; adipocyte protein aP2; adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; FABP4; fatty acid binding protein 4; Fatty acid binding protein 4 adipocyte; fatty acid binding protein 4, adipocyte; fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL S 104; HEL-S-104; Lbpl; myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.