missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4/A-FABP Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90054-50UL
This item is not returnable.
View return policy
Description
FABP4/A-FABP Monoclonal specifically detects FABP4/A-FABP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FABP4/A-FABP | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| Flow Cytometry 1:25-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
| Recombinant Human FABP4/A-FABP protein | |
| 50 μg | |
| Cancer, Phospho Specific, Stem Cell Markers | |
| 2167 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction