missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4/A-FABP Mouse anti-Human, Clone: FABP4/4422, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £415.00
Specifications
| Antigen | FABP4/A-FABP |
|---|---|
| Clone | FABP4/4422 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388816
|
Novus Biologicals
NBP3-13879-20UG |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18389903
|
Novus Biologicals
NBP3-13879-100UG |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP4/A-FABP Monoclonal antibody specifically detects FABP4/A-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| FABP4/A-FABP | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
| Recombinant fragment (around aa1-132) of human FABP4/A-FABP protein (exact sequence is proprietary) (Uniprot: P15090) | |
| Primary | |
| Store at 4°C. |
| FABP4/4422 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P15090 | |
| 2167 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title