missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4/A-FABP Antibody (rFABP4/8536), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20653-100ug
This item is not returnable.
View return policy
Description
FABP4/A-FABP Monoclonal antibody specifically detects FABP4/A-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FABP4/A-FABP | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
| Recombinant fragment (around aa1-132) of human FABP4/A-FABP protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Phospho Specific, Stem Cell Markers | |
| 2167 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rFABP4/8536 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction