missing translation for 'onlineSavingsMsg'
Learn More

FABP4/A-FABP Antibody (001), DyLight 350, Novus Biologicals™

Product Code. 18344521 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344521 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344521 Supplier Novus Biologicals Supplier No. NBP290054UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FABP4/A-FABP Monoclonal specifically detects FABP4/A-FABP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP4/A-FABP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP4/A-FABP (P15090; Cys2-Ala132,29Ala/Thr).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2167
Test Specificity Has cross-reactivity in ELISA with: Mouse FABP3 No cross-reactivity in ELISA with: Human FABP6
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.