Learn More
Invitrogen™ FABP3 (cardiac) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHFABP3X5
Additional Details : Weight : 0.33820kg
Description
The Human FABP3 (cardiac) ELISA quantitates Hu FABP3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP3. Principle of the method The Human FABP3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
MDGI /FABP3 is a small cytosolic protein responsible for the transport and deposition of fatty acids inside the cell.Specifications
P05413 | |
1.5 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
2170 | |
<12% | |
Assay Diluent Concentrate, Biotinylated Detection Antibody, Wash Buffer, Pre-coated 96 well plates, Adhesive Plate Covers, Standard, SAV-HRP, Stop Solution, Chromogen | |
5 x 96 Tests | |
Other Research | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.05 ng/mL to 200 ng/mL | |
1.5 ng/mL to 200 ng/mL | |
Biotin | |
Other Proteins | |
This ELISA pair antibody detects human FABP3. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
FABP3 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
FABP11, H-FABP, M-FABP, MDGI, O-FABP | |
1 hr. 20 min. |