missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FABP3 (cardiac) Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHFABP3

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15454983

  • £544.00 / 1 set
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

The Human FABP3 (cardiac) ELISA quantitates Hu FABP3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP3. Principle of the method The Human FABP3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

MDGI /FABP3 is a small cytosolic protein responsible for the transport and deposition of fatty acids inside the cell.
Specifications

Specifications

P05413
1.5 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
2170
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Other Research
2°C to 8°C
Human
4 hr. 45 min.
2.05-200 ng/mL
1.5ng/mL to 200ng/mL
Biotin
Other Proteins
This ELISA pair antibody detects human FABP3. Other species not determined.
Human
Colorimetric Microplate Reader
FABP3
<10%
HRP
RUO
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
FABP11, H-FABP, M-FABP, MDGI, O-FABP
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers