missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FABP2 (intestinal) ELISA Kit

Product Code. p-7231066
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15372098

Brand: Invitrogen™ EHFABP2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human FABP2 (intestinal) ELISA quantitates Hu FABP2 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP2. Principle of the method The Human FABP2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

May play roles in fatty acid transport and compartmentalization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12104
Assay Range 0.024 to 100 ng/mL
Assay Sensitivity 25 pg/mL
Calibration Range 25 pg/mL to 100 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity In this ELISA antibody pair format, no cross-reactivity was observed with rhFABP-1, 3, 5, 6, 7, 8 and 9.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2169
Gene Symbol FABP2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Other Research
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym FABPI, I-FABP
Target FABP2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.