missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP2/I-FABP Antibody (OTI2C4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70699C
This item is not returnable.
View return policy
Description
FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| FABP2/I-FABP | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FABP2 | |
| Full length recombinant protein of human FABP2/I-FABP (NP_000125) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2C4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
| Mouse | |
| Affinity Purified | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 2169.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction