missing translation for 'onlineSavingsMsg'
Learn More

FABP2/I-FABP Antibody (FABP2/7669) [Janelia Fluor™ 585], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-21004JF585

Product Code. 30171480

  • £526.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FABP2/I-FABP
Monoclonal
Janelia Fluor 585
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Recombinant full length human FABP2/I-FABP protein
0.1 mL
Cholesterol Metabolism, Lipid and Metabolism
2169
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
FABP2/7669
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.