missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Human∣Mouse samples. It is validated for Flow Cytometry∣Western Blot.
Specifications
Specifications
| Antigen | FABP1/L-FABP |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | SR1173 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human FABP1/L-FABP (Uniprot #: P07148) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?