missing translation for 'onlineSavingsMsg'
Learn More

FABP1/L-FABP Antibody (965616) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30738224 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738224 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738224 Supplier Novus Biologicals Supplier No. FAB29644MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FABP1/L-FABP Monoclonal antibody specifically detects FABP1/L-FABP in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP1/L-FABP
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 965616
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Host Species Mouse
Immunogen E. coli-derived recombinant human FABP1/L-FABP, Met1-Ile127, Accession # P07148
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2168
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.