missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAAP24 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530903

Product Code. 13455407

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (80%), Rat (80%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FAAP24 is a component of the Fanconi anemia (FA) core complex (see MIM 227650), which plays a crucial role in DNA damage response (Ciccia et al., 2007 [PubMed 17289582]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAAP24
Polyclonal
Unconjugated
FAAP24
C19orf40; FAAP24; Fanconi anemia core complex associated protein 24; Fanconi anemia core complex-associated protein 24; Fanconi anemia-associated protein of 24 kDa
Rabbit
Antigen affinity chromatography
RUO
91442
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.94 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9BTP7
FAAP24
Recombinant protein encompassing a sequence within the center region of human FAAP24. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.