missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30017
This item is not returnable.
View return policy
Description
FAAP Polyclonal antibody specifically detects FAAP in Human,Mouse samples. It is validated for Western Blot
Specifications
| FAAP | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ankyrin repeat domain 54, C22orf28, chromosome 22 open reading frame 28, DJ149A16.6, EC 6.5.1.3, HSPC117, hypothetical protein LOC51493, novel protein HSPC117, RP1-149A16.6 | |
| Between 200 and 250 | |
| 100 μg | |
| Cell Biology | |
| 51493 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction