missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAAH Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142404
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
Specifications
| FAAH | |
| Polyclonal | |
| Unconjugated | |
| FAAH | |
| amide hydrolase; anandamide amidohydrolase 1; AW412498; FA2H; Faah; FAAH1; FAAH-1; fatty acid amide hydrolase; fatty acid amide hydrolase protein; fatty-acid amide hydrolase 1; fatty-acid amide hydrolase 1-like; LOC100911581; MGC102823; MGC138146; Oleamide hydrolase; Oleamide hydrolase 1; PSAB | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2166 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O00519 | |
| FAAH | |
| Peptide with sequence C-REVERLMTPEKQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction