missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM53C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560125
This item is not returnable.
View return policy
Description
Immunogen sequence: WRPRGLRNLP RSRSQPCDLD ARKTGVKRRH EEDPRRLRPS LDFDKMNQKP YSGGLCLQET AREGSSISPP WFMACSPPPL SASCSPTGGS SQVLSESEE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
The protein encoded by this gene belongs to the FAM53 protein family. FAM53 protein family members bind to a transcriptional regulator that modulates cell proliferation. Alternative splicing results in multiple transcript variants.
Specifications
| FAM53C | |
| Polyclonal | |
| Unconjugated | |
| FAM53C | |
| 2810012G03Rik; C5orf6; FAM53C; family with sequence similarity 53 member C; family with sequence similarity 53, member C; Gm21179; Protein FAM53C; putative nuclear protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NYF3 | |
| FAM53C | |
| Recombinant protein corresponding to Human FA53C. Recombinant protein control fragment (Product #RP-100131). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction