missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FA2H Antibody (OTI2E3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72192
This item is not returnable.
View return policy
Description
FA2H Monoclonal specifically detects FA2H in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FA2H | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
| FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
| Mouse | |
| 42.6 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2E3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| FA2H | |
| Full length human recombinant protein of human FA2H(NP_077282) produced in HEK293T cell. | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 79152 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction