missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FA2H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30866-100ug
This item is not returnable.
View return policy
Description
FA2H Polyclonal antibody specifically detects FA2H in Human samples. It is validated for Western Blot
Specifications
| FA2H | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
| Synthetic peptide derived from human FA2H protein. | |
| 100 μg | |
| Lipid and Metabolism | |
| 79152 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction