missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21481-1-AP-20UL
This item is not returnable.
View return policy
Description
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.Specifications
| F9 | |
| Polyclonal | |
| Unconjugated | |
| F9 | |
| Christmas factor, coagulation factor IX, F9, FIX, HEMB, PTC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2158 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00740 | |
| F9 | |
| F9 Fusion Protein Ag15723 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction