missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Rat anti-Mouse, Clone: T45-2342, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 565409
This item is not returnable.
View return policy
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
Specifications
| F4/80 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 0.1 mg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| T45-2342 | |
| Unconjugated | |
| Gpf480; F480; Emr1; Ly71; DD7A5-7; EGF-TM7; TM7LN3 | |
| Mouse F4/80 Recombinant Protein | |
| RUO | |
| Primary | |
| The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Store undiluted at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction