missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Rat, anti-Mouse, Alexa Fluor 488, Clone: BM8, STEMCELL Technologies
Rat Monoclonal Antibody
£251.10 - £544.05
Specifications
| Antigen | F4/80 |
|---|---|
| Clone | BM8 |
| Applications | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198581
|
Stemcell Technologies
60027AD.1 |
25 μg |
£251.10
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17188581
|
Stemcell Technologies
60027AD |
100 μg |
£544.05
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The BM8 antibody reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ∼160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors
- F4/80 is considered a marker of choice for the identification of mature tissue macrophages, being broadly but variably expressed by this cell type in the liver (Kupffer cells), skin (Langerhans cells), bone marrow stroma, pancreas, thymus, spleen (red pulp), lung, and other tissues
- It is also expressed by circulating monocytes, eosinophils and a subset of dendritic cells
- F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770)
Specifications
| F4/80 | |
| Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor 488 | |
| Human, Mouse | |
| Primary |
| BM8 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title