Learn More
Invitrogen™ F4/80 Monoclonal Antibody (BM8), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 365-4801-80
Description
The BM8 MAb reacts with mouse F4/80 antigen, an app. 160 kDa surface receptor. It belongs to the EGF-TM7 family of proteins. As such it contains seven EGF-like domains on its extracellular N-terminus, seven transmembrane spanning sequences, and an intracellular C-terminal domain showing homology to other TM7 superfamily members. The F4/80 antigen is expressed by a majority of mature macrophages and is one of the best markers for this population of cells. However, other cell types, such as peritoneal eosinophils, Langerhans cells, and some other dendritic cell subtypes, have been reported to express this antigen as well. Expression of F4/80 commences during early myeloid development in vivo and can be upregulated on BM cells stimulated in vitro with M-CSF. Some populations of macrophages, especially in the lymphoid microenvironment, may be devoid of F4/80. Applications Reported: This BM8 Ab has been reported for use in flow cytometric analysis. Applications Tested: This BM8 Ab has been tested by flow cytometric analysis of mouse resident peritoneal macrophages. This may be used at less than or equal to 1ug per test. A test is defined as the amount (ug) of Ab that will stain a cell sample in a final volume of 100uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. BUV563 is a tandem dye that emits at 564nm and is intended for use on cytometers equipped with an UV (355nm) laser. Excitation: 350nm; Emission: 564nm; Laser: UV Laser.
F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Specifications
| F4/80 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q61549 | |
| ADGRE1 | |
| Affinity chromatography | |
| RUO | |
| 13733 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| BM8 | |
| Brilliant Ultraviolet 563 | |
| ADGRE1 | |
| ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.