missing translation for 'onlineSavingsMsg'
Learn More

F4/80 Antigen Rat, PE-Cy7, Clone: BM8.1, enQuireBio™

Rat Monoclonal Antibody

£165.90 - £441.00

Specifications

Antigen F4/80 Antigen
Clone BM8.1
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15912368
View Documents
EnquireBio
QAB68-PE7-25ug
25 μg
£165.90
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15902368
View Documents
EnquireBio
QAB68-PE7-100ug
100 μg
£441.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

F4/80 Antigen
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
13733
IgG2b κ
Protein A/G Purified
BM8.1
Flow Cytometry
PE-Cyanine7
RUO
XP_006523664
Adgre1
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.