missing translation for 'onlineSavingsMsg'
Learn More

F4/80 Antibody (Cl:A3-1 (recombinant version)), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18426356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426356 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426356 Supplier Novus Biologicals Supplier No. NBP281029MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen F4/80
Applications Flow Cytometry
Classification Monoclonal
Clone Cl:A3-1 (recombinant version)
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3
Host Species Rabbit
Immunogen This F4/80 Antibody (Cl:A3-1 (recombinant version)) - Chimeric was developed against Thioglycollate stimulated peritoneal macrophages of mouse origin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 13733
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.