missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antibody (Cl:A3-1 (recombinant version)), DyLight 550, Novus Biologicals™
Shop All Bio Techne ProductsDescription
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
| Antigen | F4/80 |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | Cl:A3-1 (recombinant version) |
| Conjugate | DyLight 550 |
| Dilution | Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 |
| Gene Symbols | Adgre1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?