missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antibody (Cl:A3-1 (recombinant version)), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81030CL1
This item is not returnable.
View return policy
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry
Specifications
| F4/80 | |
| Monoclonal | |
| CoraFluor 1 | |
| ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3 | |
| This F4/80 Antibody (Cl:A3-1 (recombinant version)) was developed against Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
| 0.1 mL | |
| Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity | |
| 13733 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry | |
| Cl:A3-1 (recombinant version) | |
| PBS | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction