missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen).
Specifications
Specifications
| Antigen | DYKDDDDK Epitope Tag |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | L5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) |
| Formulation | Tissue culture supernatant with 0.1% Sodium Azide |
| Gene Alias | anti-flag L5, DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag |
| Host Species | Rat |
| Immunogen | DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?