missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F1A alpha Polyclonal Antibody
GREENER_CHOICE

Product Code. 15777751 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15777751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15777751 Supplier Invitrogen™ Supplier No. PA559906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RFAQVFSQMI HLNETVKAPD IECVLRCSVL EIEQSMNRVK NISDADVHNA MDNYECNLYT FLYLVCISTK TQCSEEDQCK INKQIYNLIH Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Fas and tumor necrosis factor receptor 1 (TNFR1) are two prototype members in the death receptor family. A novel protein that associates with the intracellular domains of Fas and TNFR1 was recently identified and designated F1Aalpha and FEM1-b. F1Aalpha/FEM1-b is the homologue of C. elegans sex determining protein FEM-1. FEM-1/F1Aalpha is cleaved by CED-3 and caspase. FEM-1/F1Aalpha associates with CED-4 and its mammalian homologue Apaf-1. Overexpression of F1Aalpha induces apoptosis. F1Aalpha is therefore a novel member of the death receptor associated protein that mediates apoptosis. F1Aalpha is expressed in a variety of human and mouse tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen F1A alpha
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FEM1B
Gene Accession No. Q9UK73
Gene Alias DKFZp451E0710; F1aa; F1A-alpha; fem-1 homolog B; fem-1 homolog b (C. elegans); Fem1b; FEM1-beta; FEM-1-like death receptor binding protein; fem-1-like death receptor-binding protein alpha; Fem-1-like in apoptotic pathway protein alpha; feminization 1 homolog b; feminization 1 homolog b (C. elegans); FIAA; Kiaa0396; mKIAA0396; mt-Fem; Protein fem-1 homolog B
Gene Symbols FEM1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human F1A alpha. Recombinant protein control fragment (Product #RP-98129).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.