missing translation for 'onlineSavingsMsg'
Learn More

F16, MedChemExpress

Product Code. 16452988
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16452988

Brand: Medchem Express HY100395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress F16 is a potent growth inhibitor of the neu-overexpressing cells and also selectively inhibits proliferation of mammary epithelial as well as a variety of mouse mammary tumor and human breast cancer cell lines.

F16 is a potent growth inhibitor of the neu-overexpressing cells and also selectively inhibits proliferation of mammary epithelial as well as a variety of mouse mammary tumor and human breast cancer cell lines.

Specifications

Chemical Name or Material F16
Physical Form Solid
CAS 36098-33-6
Color Orange
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H15IN2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 31 mg/mL (85.59 mM) ∣Ethanol : 1 mg/mL (2.76 mM; Need ultrasonic)
SMILES C[N+]1=CC=C(/C=C/C2=CNC3=C2C=CC=C3)C=C1.[I-]
Molecular Weight (g/mol) 362.21
Formula Weight 362.21
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.