missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F16, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Description
F16 is a potent growth inhibitor of the neu-overexpressing cells and also selectively inhibits proliferation of mammary epithelial as well as a variety of mouse mammary tumor and human breast cancer cell lines.
Specifications
Specifications
| Chemical Name or Material | F16 |
| Physical Form | Solid |
| CAS | 36098-33-6 |
| Color | Orange |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H15IN2 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 31 mg/mL (85.59 mM) ∣Ethanol : 1 mg/mL (2.76 mM; Need ultrasonic) |
| SMILES | C[N+]1=CC=C(/C=C/C2=CNC3=C2C=CC=C3)C=C1.[I-] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction