missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F131C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556623
This item is not returnable.
View return policy
Description
Immunogen sequence: AEDEHYCCLP DELREARFAA GVAEQFAITE ATLSAWSSLD EEELHPENSP QGIVQLQDLE SIYLQDSLPS G Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
F131C gene encodes a member of the superoxide dismutase (SOD) protein family. SODs are antioxidant enzymes that catalyze the conversion of superoxide radicals into hydrogen peroxide and oxygen, which may protect the brain, lungs, and other tissues from oxidative stress. Proteolytic processing of the encoded protein results in the formation of two distinct homotetramers that differ in their ability to interact with the extracellular matrix (ECM). Homotetramers consisting of the intact protein, or type C subunit, exhibit high affinity for heparin and are anchored to the ECM. Homotetramers consisting of a proteolytically cleaved form of the protein, or type A subunit, exhibit low affinity for heparin and do not interact with the ECM. A mutation in this gene may be associated with increased heart disease risk.
Specifications
| F131C | |
| Polyclonal | |
| Unconjugated | |
| FAM131C | |
| C1orf117; FAM131C; family with sequence similarity 131 member C; family with sequence similarity 131, member C; Gm693; hypothetical protein LOC619198; protein FAM131C; RP11-5P18.9; zgc:110264 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 348487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96AQ9 | |
| FAM131C | |
| Recombinant protein corresponding to Human F131C. Recombinant protein control fragment (Product #RP-94504). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction