Learn More
F11R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26941-1-AP-150UL
This item is not returnable.
View return policy
Description
The F11R antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human F11R. This antibody recognizes human antigen. The F11R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| F11R | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 50848 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 433 μg/mL | |
| Rabbit | |
| Primary | |
| Human |