missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F-Spondin/SPON1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | F-Spondin/SPON1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18324066
|
Novus Biologicals
NBP3-03648-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323937
|
Novus Biologicals
NBP3-03648-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
F-Spondin/SPON1 Polyclonal specifically detects F-Spondin/SPON1 in Mouse samples. It is validated for Western Blot.Specifications
| F-Spondin/SPON1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| f-spondin, KIAA0762VSGP/F-spondin, MGC10724, spondin 1, (f-spondin) extracellular matrix protein, spondin 1, extracellular matrix protein, spondin-1, Vascular smooth muscle cell growth-promoting factor, VSGP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-300 of human F-Spondin/SPON1 (NP_006099.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 10418 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title