missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F box protein 38 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | F box protein 38 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348626
|
Novus Biologicals
NBP3-03841-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303176
|
Novus Biologicals
NBP3-03841-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
F box protein 38 Polyclonal specifically detects F box protein 38 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| F box protein 38 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 81545 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| F-box only protein 38, F-box protein 38, FBX38, FLJ13962, Modulator of KLF7 activity homolog, MoKA, MOKAFbx38, SP329 | |
| Recombinant protein of human F box protein 38. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title