missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F-box protein 15/FBXO15 Antibody (612834) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6035X
This item is not returnable.
View return policy
Description
F-box protein 15/FBXO15 Monoclonal antibody specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| F-box protein 15/FBXO15 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 201456 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 612834 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| F-box only protein 15, F-box protein 15, FBX15MGC39671 | |
| E. coli-derived recombinant human F-box protein 15/FBXO15 isoform 2,, Asn298-Tyr434, Accession # Q8NCQ5-2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction