missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One F-Bottom 96-well Assay Microplates
GREENER_CHOICE

Product Code. 10365532
Change view
Click to view available options
Color:
Black
Clear
White
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
Pack of 120
Pack of 32
Pack of 40
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Lid unitSize
10365532 White Non-sterile Without Lid Pack of 40
10284272 White Sterile With Lid Pack of 32
10164911 White Sterile Without Lid Pack of 40
11827822 Clear Non-sterile With Lid Pack of 120
10341341 Clear Sterile Without Lid Pack of 100
10177050 Clear Sterile With Lid Pack of 100
10033211 Black Non-sterile Without Lid Pack of 40
10214232 Black Sterile Without Lid Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10365532 Supplier Greiner Bio-One Supplier No. 655075
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Features a chimney-like arrangement of the wells that allows each well to stand on its own. Greiner Bio One™ F-Bottom 96-Well Assay Microplates are constructed of polystyrene, a highly clear polymer with excellent optical properties making it ideal for precise optical measurements. Polystyrene is also characterized by its ability to bind biomolecules, such as proteins and is suitable for work with cell cultures.

  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
  • Manufactured from raw materials tested for leachables
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN ISO 9001
Certified RNase-/D-Nase, human DNA free
Format Standard
Lid Without Lid
Culture Area 34 mm2
Array 8 x 12
Color White
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 34 μL
Sterility Non-sterile
Surface Treatment LUMITRAC 200
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. Heat Stability -20 to +60°C
Max. RCF 4800 x g: Swinging-Bucket Rotor
Quantity 40/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Height (Metric) 11.5 mm
Diameter (Metric) Well 5 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.