missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F-actin Monoclonal Antibody (NH3)

Mouse Monoclonal Antibody
£337.00 - £562.00
Specifications
| Antigen | F-actin |
|---|---|
| Clone | NH3 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11515412
|
Invitrogen™
MA180163 |
50 μg |
£337.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11575522
![]() |
Invitrogen™
MA180729 |
500 μg |
£562.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse anti Human F-Actin antibody, clone NH3 recognizes human Filamentous actin (F-actin).
This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins.Specifications
| F-actin | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P29751, P60709, P60710, P60711 | |
| 100009272, 11461, 60, 81822 | |
| Human monocytes and U937 cell line. | |
| Antibody |
| NH3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgM | |
| Human, Mouse, Rat, Rabbit | |
| ACTB | |
| ACT; ACT-5; ACTB; ACTG; ACTG1; actin beta; actin gamma 1; actin, beta; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actx; Atcb; beta cytoskeletal actin; beta-actin; BRWS1; BRWS2; cytoplasmic beta-actin; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E430023M04Rik; epididymis luminal protein 176; gamma non-muscle actin; Gamma-actin; HEL-176; PS1TP5-binding protein 1; PS1TP5BP1 | |
| ACTB | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title