Learn More
Invitrogen™ F(ab)-Goat anti-Rabbit IgG Fc Secondary Antibody, Alexa Fluor™ 555
F(ab)-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ A66786
Description
This F(ab)-Goat anti-Rabbit IgG Fc specific antibody product is a goat polyclonal antibody F(ab) fragment that is specific to the Fc region of rabbit IgG. The product has minimal to no cross-reactivity towards polyclonal mouse, rat, guinea pig, bovine, hamster, horse, sheep, human, monkey, donkey IgG, and chicken IgY as tested by direct ELISA. Cross-reactivity towards other immunoglobulins has not been tested. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. F(ab) antibody fragment offer several benefits over whole IgGs, due to their smaller size (~50 kD vs. ~150 kD) and the absence of the Fc region. The smaller size is expected to allow better penetration into tissue sections and improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells/ tissues with Fc receptors (such as immune cells) and other Fc interaction-related interference. Fc specific F(ab) fragments provide a range of multiplexing options beyond what is possible with whole IgGs for applications such as immunocytochemistry, immunohistochemistry, flow cytometry, and western blotting. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG Fc | |
| Polyclonal | |
| Alexa Fluor 555 | |
| Goat | |
| Size-exclusion chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG F(ab) |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Rabbit IgG Fc. | |
| 50 μg | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.