Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 705
F(ab')2-Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ Q11461MP
Product Code. 10632834
  • £450.00 / 200µL
For Research Use Only. All usage must comply with product instructions.
Description
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit IgG (H+L)
Polyclonal
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3
Gamma Immunoglobins Heavy and Light chains.
200 μL
Secondary
4°C, store in dark
Liquid
Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Qdot 705
Goat
Purified
RUO
Rabbit
Secondary Antibody