missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, PE-Cyanine5.5
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ L43018
This item is not returnable.
View return policy
Description
F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody for Flow
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG (H+L) | |
| Polyclonal | |
| PBS with 0.1% sodium azide | |
| Gamma Immunoglobin. | |
| 500 μL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry | |
| PE-Cyanine5.5 | |
| Goat | |
| Purified | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto