Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, NANOGOLD™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ N24916
Description
The NANOGOLD conjugate of Fab' fragment of goat anti-rabbit IgG is labeled with small uniformly sized (1.4 nm) gold particles at a ~1:1 ratio in the hinge region, remote from the antigen-binding site and can be used in immunoblotting, light microscopy, and electron microscopy applications.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG (H+L) | |
| Polyclonal | |
| Nanogold | |
| Goat | |
| Purified | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
| Immunomicroscopy, Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% BSA and 0.05% sodium azide; pH 7.4 | |
| Gamma Immunoglobin. | |
| 1 mL | |
| Secondary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.