Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, APC
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31984 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31984 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, mouse, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 73 at 650 nm for a 1% solution for only those APC molecules to which at least one molecule of active antibody is bound) Allophycocyanin Source: Spirulina platensis Store product protected from light at 4°C until opened. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. Do not freeze. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Allophycocyanin Amax= 650 nm; Emax= 660 nm. Reconstitute with 0.5 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.