Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 655, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen Q11021MP
Code : 50
Additional Details : Weight : 0.50000kg
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Mouse IgG (H+L) | |
Polyclonal | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Gamma Immunoglobins Heavy and Light chains | |
200 μL | |
Secondary | |
4° C, store in dark | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Qdot 655 | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |